Learn More
Ethos Biosciences Mayer's Hematoxylin, 5gal, Ethos
Supplier:  Ethos Biosciences 70215G
Encompass
Mayer’s Hematoxylin is a nuclear staining solution designed for histology applications. Formulated as a progressive stain, it provides clear, crisp nuclear detail without the need for differentiation steps. The gentle staining action makes it ideal for delicate tissues and routine procedures, including H&E and special staining protocols. The formulation delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Mayer’s Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74277