Learn More
Ethos Biosciences Gill 3X Hematoxylin, 16oz, Ethos
Supplier:  Ethos Biosciences 701416
Encompass_Preferred
Gill 3X Hematoxylin is a nuclear staining solution designed for histology applications. Formulated for progressive staining, it provides intense, crisp nuclear detail without the need for differentiation steps. The 3X formulation offers the highest staining strength in the Gill series, making it suitable for thicker sections and applications requiring deeper nuclear contrast. It delivers consistent color development for reliable microscopic evaluation and reproducible results. Suitable for clinical, research, and educational laboratories, Gill 3X Hematoxylin supports efficient and dependable staining workflows. Supplied in chemically compatible containers for convenient laboratory use.
Catalog No. 50-005-74264