Learn More
Electron Microscopy Sciences Stainless Steel EMS Staining Tray | 60 Slides
SDP
Supplier:  Electron Microscopy Sciences 7222006
Encompass_Preferred
These stainless steel slide staining trays are ideal for routine and special histology staining procedures. The staining trays have removable handles. Accepts 3" W (76 mm) x 1", 1¿", or 2" H slides.
Capacity: 60 slides
Catalog No. 50-193-1274