Learn More
Sigma Aldrich Fine Chemicals Biosciences Stainless steel 304 syring1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1011331EA
Encompass
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved deflecting tip to prevent coring during needle insertion through the septum.Standard Hub with 304 Stainless Steel Tubing
Catalog No. 50-290-0962