Learn More
TA Instruments Stainless Steel, 50 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Supplier:  TA Instruments 402622.901
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Quick Change Plate (50 mm) is designed for use with the ARES-G2 APS Standard Flat Plate (P/N: 402619.901). Ensures accurate positioning and reproducible measurement results across HR X0, DHR, and ARES-G2 platforms.
Catalog No. 50-005-80721