Learn More
Braintree Scientific Stainless Steel Brain Matrice, Rat 175-300gr, 0.5 mm Coronal
SDP
Supplier:  Braintree Scientific BSSS605C
Encompass
Stainless Steel Brain Matrices These highly polished high quality stainless steel matrices are precisely machined to ensure reproducible sections. The brain matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1mm or 0.5mm intervals. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4421