Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 9951 (5 Canisters)
SDP
Supplier:  Alkali Scientific FC4R99515
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 5 BAXTER/FENWAL C 4R9951 Aluminum Canisters Blood Bag Number: 4R 9951 Dimensions (D x H x L): 5 11/6 x 18 3/4 x 9/16″ 145 x 476 x 15mm Canister: C 4R9951 Platform Dividers P20 4R9951 P24 4R9951 P31 4R9951 P39 4R9951
Catalog No. 50-006-62510