Learn More
Alkali Scientific Stainless Steel Canister Frames for CHARTERMED Blood Bag # 7400 2, 7403 2, 7410 2 & 7025 2 (3 Canisters)
SDP
Supplier:  Alkali Scientific FC74003
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 3 CHARTERMED C 7400 Aluminum Canisters Blood Bag Number: 7400 2 7403 2 7410 2 7025 2 Dimensions (D x H x L): 8 1/4 x 27 13/16 x 9/16″ 209 x 707 x 15mm Canister: C 7400 Platform Dividers P20 7400 P24 7400 P31 7400 P39 7400
Catalog No. 50-006-62022