Learn More
TA Instruments Stainless Steel, Cross Hatched, 40 mm Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Supplier:  TA Instruments 517400.945
Encompass_Preferred
HR X0 Stainless Steel Cross Hatched Plate (40 mm) is an upper plate compatible with lower Peltier plates. It uses Smart Swap 2 geometry and 1 mm deep, 1 mm peak-to-peak cross hatching to provide uniform sample grip and precise measurements on HR X0 instruments.
Catalog No. 50-005-80727