Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for 2" Boxes, Vertical Configuration, 11 Box Capacity
SDP
Supplier:  Alkali Scientific CF112
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Vertical Racks for Chest Freezers and Liquid Nitrogen Tanks are made of corrosion resistant stainless steel. Racks are made for standard 2" boxes. All racks include stainless steel locking rods. Dimensions H x W x D: 24 1/2 x 5 5/8 x 5 1/2 inch (622 x 143 x 140mm)Compartment Dimensions h x w x d: 2 3/16 x 5 5/16 x 5 3/8 inch (55 x 134.5 x 136mm)
Catalog No. 50-302-6405