Learn More
Alkali Scientific Stainless Steel Freezer Racks
SDP
Supplier:  Alkali Scientific CF42
Encompass
The Stainless Steel Freezer Racks for 2in Boxes in a vertical configuration from Southern Labwares provide a robust and reliable storage solution for ultra-low temperature environments These racks are expertly crafted from high-quality corrosion-resistant stainless steel ensuring durability and longevity even in the demanding conditions of ultra-low (-80C) freezers Designed with versatility in mind they are ideal for both chest freezers and liquid nitrogen tanks providing a secure and organized storage option for standard 2in cardboard plastic or metal boxes Key Features High-Quality Construction Made from corrosion-resistant stainless steel these racks are built to withstand the harsh conditions of ultra-low temperature storage ensuring long-term reliability and performance Universal Compatibility Thoughtfully configured to fit all major ultra-low (-80C) freezers on the market these racks are designed for ultra-low upright freezers offering broad applicability across various labor
Catalog No. 50-302-6485