Learn More
Alkali Scientific Alkali Scientific™ Vertical Rack for 50-Cell 1.5 mL Microtube Boxes, Holds 22 boxes
SDP
Supplier:  Alkali Scientific CFM1115
Encompass
For 50-Cell 1.5 ml Storage Boxes Dimensions H x D x W: 25 9/16 x 5 7/8 x 6 1/4 inch (649 x 149 x 159mm)Compartment Size h x d x w: 2 5/16 x 5 1/2 x 5 15/16 (58 x 140 x 150.7mm) *Security lock device is ordered separately.
Catalog No. 50-302-9328