Learn More
Alkali Scientific Stainless Steel Freezer Racks
SDP
Supplier:  Alkali Scientific CFH5010
Encompass
The Vertical Freezer Racks for 50 Cell Hinge Lid Storage Boxes are designed to provide a secure and efficient storage solution for laboratories looking to organize and preserve samples in 50 cell hinge lid storage boxes within vertical freezer environments These racks are tailored to enhance space utilization facilitate easy access and ensure the longevity and integrity of stored samples Product Features Optimized for 50 Cell Hinge Lid Boxes These freezer racks are specifically engineered to accommodate 100 cell hinge lid storage boxes ensuring a snug and secure fit This design minimizes movement and provides optimal protection for the samples stored within Enhanced Space Efficiency The vertical orientation of the racks maximizes the use of vertical space within freezers significantly increasing storage capacity without sacrificing accessibility This allows laboratories to store a greater number of samples in a limited space improving organization and efficiency Robust Constructi
Catalog No. 50-302-9372