Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Freezer Racks for 3" Boxes, 4 x 5 Configuration, 20 Box Capacity
SDP
Supplier:  Alkali Scientific UFR453
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 5 Tall= Holds 20 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 15 9/16 inches (564 x 140 x 395mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (136 x 138 x 78mm)
Catalog No. 50-302-7737