Learn More
Alkali Scientific Stainless Steel Freezer Racks
SDP
Supplier:  Alkali Scientific UFDR323
Encompass
Stainless Steel Freezer Racks with Drawers for 3in Boxes 3 x 2 Configuration 6 Box Capacity Constructed of corrosion resistant stainless steel Racks fit standard 3in cardboard plastic or metal boxes Security lock devices available for each rack 3 Long x 2 Tall Holds 6 Boxes Freezer Storage Racks are constructed of high-quality corrosion resistant stainless steel Handles at both ends accommodate freezers with right or left-hand doors Our freezer racks have been configured to fit all major ultra-low (-80) freezers on the market racks are designed for ultra-low upright freezers Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer Security lock devices are also available for each rack though ordered separately Dimensions (DxWxH) 16 3/4 x 5 1/2 x 6 7/16 inches (426 x 140 x 164mm)Compartment Dimensions 5 3/8 x 5 1/4 x 3 inc
Catalog No. 50-302-8029