Learn More
TA Instruments Stainless Steel Quick Change Plate, 8 mm, HR X0, DHR, ARES-G2, APS
Supplier:  TA Instruments 402625.901
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel 8 mm Quick Change Plate is for use with ARES-G2 APS Standard Flat Plate (P/N: 402619.901).
Catalog No. 50-005-80723