Learn More
Science Company STAINLESS STEEL SCOOPULA 6IN
SDP
Supplier:  Science Company NC0829
Encompass
This item has a minimum qty of 12 per supplier requirements.
This six inch stainless steel Scoopula is ideal for handling small amounts of material in the lab and field. Use a scoopula to easily remove loose chemicals from small bottles with minimal spillage. Also good for breaking up clumped material.
Catalog No. NC0124317