Learn More
Anton Paar STAINLESS STEEL, LUER ADAPTER, FOR PELTIER, 100 mm for MCP 100
Supplier:  Anton Paar 143946
Encompass_Preferred
100 mm stainless steel sample cell (polarimeter tube) with wireless Toolmaster function which transfers temperature and path length to the MCP 100.Filling volume approximately 2 mL.Connection to a syringe through Luer filling ports.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306
Catalog No. NC1818309