Learn More
GUARDIAN EQUIPMENT Eyewash, Deck Mounted, 90º Swivel, All-Stainless Steel, Right Hand Mounting
Supplier:  GUARDIAN EQUIPMENT G1893
Encompass_Preferred
All-stainless steel eyewash for mounting on counter next to sink. Unit is constructed entirely in stainless steel. Ideal for use in highly corrosive environments, cleanrooms and pharmaceutical applications. Spray heads swivel 90 degrees from storage to operational position. Water flow is activated by flag handle.
Catalog No. NC2941335