Learn More
Alkali Scientific Alkali Scientific™ Stainless-Steel Upright Freezer Rack for 25-Place Slide Boxes, 5x4 32 Box Capacity
SDP
Supplier:  Alkali Scientific UFS2554
Encompass
For 25- Place Slide Boxes The UFS-25-54 standard upright freezer racks for storage of 25-place slide boxes, features 5 boxes deep by 4 boxes high configuration for a total storage of 20 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with 25-place slide boxes. Dimensions (DxWxH): 19 x 5 11/16 x 6 3/16 inches (483 x 145 x 156.5mm)Compartment Dimensions: 3 5/8 x 5 5/8 x 1 3/8 inches (92 x 144 x 35mm)
Catalog No. 50-302-7886