Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel Upright Freezer Rack for Plasma Boxes, 3x4 configuration, 12 Box Capacity
SDP
Supplier:  Alkali Scientific UFP34
Encompass
For Plasma Boxes Our freezer racks feature pharmaceutical grade stainless steel, double welds for long life and smooth edges for your safety and comfort.  Made of corrosion resistant stainless steel Locking rods are included Dimension L x H x W: 22 3/8 x 8 13/16 x 5 1/2 inch (568 x 224 x 140mm)Compartment Dimension l x h x w: 7 3/16 x 2 3/16 x 5 3/8 inch ( 183 x 54.9 x 136.7mm)
Catalog No. 50-302-6390