Learn More
Waters Corp Waters Corporation Needle, FEP/Stainless Steel, 30 μL
Supplier:  Waters Corp 205000369
Encompass_Preferred
This is an optional 30μL FEP/Stainless Steel Tipped needle for the ACQUITY UPLC Sample Manager.
  • Optional 30μL FEP/Stainless Steel Tipped needle.
  • Designed for ACQUITY UPLC Sample Manager.
  • Compatible with UPLC system type.
  • Suitable for ACQUITY Arc Premier System.
  • Annual replacement recommended.
  • Functions as an ACQUITY UPLC Column Manager component.
Catalog No. NC1682950