Learn More
Medchemexpress LLC Stampidine | 217178-62-6 | 100.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY122470100MG
Encompass_Preferred
Stampidine is a nucleoside reverse transcriptase inhibitor (NRTI) used in research to evaluate anti-HIV activity and resistance, exhibiting low-nanomolar potency against laboratory and clinical HIV-1 isolates.
  • Nucleoside reverse transcriptase inhibitor used in antiviral research.
  • Active at low-nanomolar IC50 against laboratory and clinical HIV-1 isolates.
  • Shows activity against some NRTI- and NNRTI-resistant strains.
  • Available as a solid and as a 10 mM solution in DMSO for assay use.
  • High reported purity suitable for research applications.
Catalog No. 50-002-30294