Learn More
Medchemexpress LLC Stampidine | 217178-62-6 | 100.0% | 50 MG

Supplier: Medchemexpress LLC HY12247050MG
Stampidine is a nucleoside reverse transcriptase inhibitor (NRTI) research compound with potent, broad-spectrum anti-HIV activity. It exhibits low-nanomolar inhibition of laboratory and clinical HIV-1 isolates and has activity against certain drug-resistant strains.
- High purity suitable for research use (99.96%).
- Broad-spectrum antiviral activity against laboratory and clinical HIV-1 isolates.
- Retains activity against some NRTI- and NNRTI-resistant strains.
- Available in small quantities for in vitro and preclinical studies.
- Molecular weight 544.29 g/mol; formula C20H23BrN3O8P.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.