Learn More
Cole-Parmer Wiremold/Legrand UL207BC Standard 6-Outlet Power Strip with 6 Ft Cord
Supplier:  Cole-Parmer 2687000
Encompass_Preferred
Each model features an aluminum housing and impact-resistant plastic end caps. All outlet strips meet UL standard 1363 and have a built-in circuit breaker. Standard outlet strips are ideal for any general-purpose indoor use. Mounting clips are included for easy installation. Units have a combination ON/OFF switch/pilot light. CSA-approved.
Catalog No. NC0872483
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov