Learn More
TA Instruments Standard Aluminum Hermetic Lids, Use with Standard Aluminum Hermetic Pans, Hermetic Seal Mode, Secure Hermetic Sealing, DSC and TGA Applications, Requires Tzero Sample Press or Standard Press, 200 Pack
Supplier:  TA Instruments 901697.901
Encompass_Preferred
Waters TA Instruments Standard Aluminum Hermetic Lids are for use with Standard Aluminum Hermetic Pans in hermetic seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. They provide secure, hermetic sealing for DSC and TGA applications.
Catalog No. 50-005-80591