Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Particle Size Standard Microsphere 3.0 µm 15 ML
SDP

Supplier:  Electron Microscopy Sciences 70896

Encompass

Polystyrene sphere particle-size standards are certified for mean diameter and are traceable to the National Bureau of Standards (NBS). The most uniform spheres available and are recommended for the highest level of measurement accuracy in the 1 to 40 micron size.
Certified Mean diameter: 3.0 µm
Nominal Mean diameter: 3.063+/-0.027
Size Uniformity std. dev. & C.V: 0.03 µm (1.0%)
Solids content: 1%

Catalog No. 50-930-526


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.