Learn More
Research Products International Corp Standard Pipette Tips
SDP
Supplier:  Research Products International Corp 141119
Encompass
Our precision molded pipet tips provide optimum fit and sealing with all major brands of single and multi-channel pipettors. Tips are constructed from low adhesion polypropylene with a flash free orifice and smooth wall surface. These features minimize sample retention to provide the highest accuracy and reproducibility. Advanced manufacturing techniques eliminates the use of chemical mold release agents and ensures a contamination free product.
Catalog No. 50-272-2985