Learn More
Perkin Elmer US LLC Standard PVC Two-Stop Peristaltic Pump Tubing - 1.85 mm I.D., Green/Green, Pkg. 12
Supplier:  Perkin Elmer US LLC N0777518
Encompass_Preferred
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-210-5280