Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Staphylococcus aureus (ATCC BAA-3336) Research Strain - Culture for Microbiology and Genomic Study

Supplier:  ATCC BAA3336

Encompass

A characterized Staphylococcus aureus culture supplied for research and genomic analysis. The strain includes genome sequence data and antibiotic susceptibility profiles to support microbiology, antimicrobial testing, and quality control workflows.

  • Organism: Staphylococcus aureus.
  • Strain: clinical isolate from decubitus.
  • Biosafety level: 2.
  • Genome length: 2,832,812 nt.
  • Typical applications: antimicrobial susceptibility testing, genomic analysis, microbiology research.
  • Storage and handling: follow supplier instructions and institutional biosafety procedures.

Catalog No. NC3538704


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.