Learn More
ATCC Staphylococcus aureus; Strain 7031
Supplier:  ATCC BAA1764
Encompass
Staphylococcus aureus strain 7031 is a whole-genome sequenced bacterial strain that has applications in antimicrobial resistance research, drug development, and infectious disease research. This drug-resistant strain was confirmed to be methicillin resistant.
Catalog No. 50-238-3696