Learn More
ATCC Staphylococcus aureus subsp. aureus Rosenbach
Supplier:  ATCC 43300/DEL
Encompass
Resistant to methicillin and oxacillin. Used in susceptibility testing, and evaluation of Mueller-Hinton Agar.
Catalog No. 50-189-157FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product