Learn More
ATCC Staphylococcus aureus; subsp. aureus; Strain Mu3
Supplier:  ATCC 700698
Encompass
Resistant to methicillin (MRSA). This strain is cited as showing heterogeneous susceptibility to vancomycin (Hiramatsu et al., 1997).
Catalog No. 50-238-3214