Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC BAA976
Encompass
Staphylococcus aureus subsp. aureus strain UT 25 is a clinical specimen isolated in 2003 from tracheal aspirate. This strain is the negative control for the inducible clindamycin-resistant “D” test.
Catalog No. 50-238-2732