Learn More
ATCC Staphylococcus aureus; subsp. aureus; UT 32
Supplier:  ATCC BAA977
Encompass
Staphylococcus aureus subsp. aureus strain UT 32 is a clinical specimen that was isolated in 2003 from a wound infection. This strain is the positive control for the inducible clindamycin-resistant “D” test.
Catalog No. 50-238-3117