Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC BAA1556
Encompass
Staphylococcus aureus subsp. aureus strain FPR3757 was isolated from the wrist abscess of an HIV+ man with a history of IV drug use. This whole-genome sequenced bacterial culture is resistant to methicillin and oxacillin and can be used in drug discovery.
Catalog No. 50-238-2369
 Restricted Item