Learn More
ATCC Saccharopolyspora spinosa; Strain A83543.1
Supplier:  ATCC 49460
Encompass
Saccharopolyspora spinosa strain A83543.1 is a whole-genome sequenced bacterial type strain that was isolated from the soil of a sugar-mill rum still in the Virgin Islands.
Catalog No. 50-238-1520