Learn More
Genetex Inc StAR Monoclonal Antibody [HMV306] HistoMAX™ (GTX639930) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639930
Encompass_Preferred
The StAR antibody [HMV306] HistoMAX™ (GTX639930) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human follicular cyst from ovary tissue using GTX639930 StAR antibody [HMV306] HistoMAX. Strong cytoplasmic StAR staining of theca interna cells of a follicular cyst.
Catalog No. NC3669300