Learn More
Genetex Inc StAR Monoclonal Antibody [MSVA-740R] HistoMAX™ (GTX04483) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04483
Encompass_Preferred
The StAR antibody [MSVA-740R] HistoMAX™ (GTX04483) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human testis tissue using GTX04483 StAR antibody [MSVA-740R] HistoMAX. Strong cytoplasmic StAR immunostaining of Leydig cells.
Catalog No. NC3667795