Learn More
ABBERIOR INSTRUMENTS AMERICA LLC STAR ORANGE goat anti-mouse IgG
Supplier:  ABBERIOR INSTRUMENTS AMERICA LLC STORANGE1001500UG
Encompass_Preferred
abberior STAR ORANGE is a novel fluorescent dye developed for STED and confocal microscopy in the orange spectral region. Introducing negatively charged groups into its molecular structure making the dye excellently water soluble and enables background-free imaging. Thus, abberior STAR ORANGE is highly photostable and a bright orange fluorescent dye, which can be effectively excited with excitation light between 550 - 610 nm. For STED microscopy, this dye can be most efficiently used with a STED laser wavelength between 750 - 800 nm. Our abberior STAR ORANGE can substitute dyes like ATTO 594 or Alexa Fluor 594. Together with our abberior STAR RED you can obtain stunning 2 color STED results. Organic fluorescent dye conjugated with polyclonal secondary anti-rabbit IgG antibody, host: goat, concentration 1 mg/ml. For scientific use only. Must not be used for diagnostic or medical purposes.
Catalog No. NC1933863