Learn More
Medchemexpress LLC Starburst 1st generation (13.93% w/w in water) | 142986-44-5 | 13.93% w/w in water | 1429.85 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16947450MG
Encompass_Preferred
Starburst 1st Generation (PAMAM G1.0) is a Polyamidoamine dendrimer with amine termini. It has been utilized as a drug delivery system in vitro.
  • Used as a drug delivery system in vitro.
  • Increases the solubility of 5-Fluorouracil when conjugated to di-n-dodecylamine and encapsulating 5-Fluorouracil.
  • Cytotoxic to AGS gastric adenocarcinoma cells when used in the aforementioned conjugated and encapsulating form.
Catalog No. 50-003-90393