Learn More
MSC Starrett Blade, 12 in, Vernier Protractor
Supplier: MSC 58699828
The Starrett PT04781 Blade is designed for use with C359/C364 series bevel protractors. This 12 inch blade is suitable for precision angle measurements.
- Designed for C359/C364 series bevel protractors.
- Features a 12 inch blade length.
- Suitable for vernier protractor types.
- Manufactured in the United States with global content.