Learn More
MSC Starrett Combination Square, 4 in Blade, Square Head, 4R Graduation
Supplier:  MSC 86402138
Encompass
This combination square with a square head features a reversible lock bolt and a hardened steel, photo-engraved blade with a satin chrome finish. The square heads are made of forged hardened steel with a smooth, black enamel finish, and the 4 inch blade features 4R graduations.
  • Reversible lock bolt
  • Hardened steel, photo-engraved blade
  • Satin chrome finish blade
  • Forged and hardened steel heads with smooth black enamel finish
  • 4R graduations for precise measurements
Catalog No. 50-005-31333