Learn More
MSC Starrett Combination Square Blade, 12 inch, 4R Graduation, Steel, Satin Chrome
Supplier:  MSC 86401098
Encompass
This combination square blade is designed for precise measurements in various applications. Made from durable steel with a satin chrome finish, it provides clear and accurate readings for professionals.
  • Made from steel for durability.
  • Features a satin chrome finish for clear visibility and corrosion resistance.
  • Equipped with 4R graduations, including 8ths, 16ths, quick reading 32nds, and 64ths.
  • Measures 12 inches in length.
  • Has a blade width of 1 "
  • Features a blade thickness of 3/32 "
  • Manufactured in the United States.
Catalog No. 50-005-27236