Learn More
MSC Starrett Dial Test Indicator, Swivel Head, Black Dial, .030 in/.0005 in
Supplier: MSC 86435781
The Starrett B811-5CZ Dial Test Indicator is a versatile measuring tool designed for precision applications. It features a swivel head for flexible positioning and a black dial for enhanced readability. This indicator provides a .030 inch measuring range with .0005 inch graduations, making it suitable for detailed measurements in various orientations.
- Automatic reversing contact point maintains clockwise hand rotation.
- Frictionally adjustable and replaceable contacts.
- Smooth, jeweled movement for reliable operation.
- Large 1-3/8 inch (35 mm) dial diameter for increased readability.
- Swivel head allows positioning from horizontal to vertical and at any angle up to 90 degrees.
- Two positioning mounts compatible with dovetail test indicator accessories.
- Manufactured in the USA.