Learn More
MSC Starrett Gage Block, Rectangular, Steel, .900 Inch, Grade 0
Supplier: MSC 08470437
This precision gage block serves as a primary standard for dimensional quality control in manufacturing. Essential characteristics for these blocks include accuracy, surface finish, wear resistance, and dimensional stability. Made from high-grade steel, this rectangular gage block is suitable for shop floor environments and the calibration of measuring equipment.
- Accuracy meets or exceeds known specifications, with excellent flatness, parallelism, and surface finish.
- Maintains size over time, changing only through normal wear.
- Steel blocks have a Rockwell "C" hardness of approximately 64-65.
- Constructed from high-grade steel, suitable for shop floor use.
- B89.1.9 Accuracy Grade is 0.
- Nominal size of .900 in.
- Designed for inspection applications.