Learn More
MSC Starrett Radius Gage, 3/32 Inch, Stainless Steel, Satin Chrome Finish
Supplier:  MSC 58696071
Encompass
This radius gage is designed for easier, faster, and more accurate checking or laying out of radii or fillets. It is constructed from durable stainless steel and features a satin chrome finish.
  • Designed for easier, faster, and more accurate checking or laying out of radii or fillets.
  • Made of stainless steel.
  • Features a satin chrome finish.
  • Configured with convex and concave radii per each leaf.
Catalog No. 50-005-31733