Learn More
Medchemexpress LLC STAT3-D11-PROTAC-VHL | 96.3% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1727671MG
Encompass_Preferred
This PROTAC degrader targets Signal Transducer and Activator of Transcription 3 (STAT3). It exhibits anti-tumor activity with IC50 values of 1335 nM and 1973 nM against HeLa and MCF-7 cells, respectively. The compound binds to the DNA-binding domain of STAT3, recruits the E3 ligase VHL to form a ternary complex, leading to ubiquitination and subsequent degradation by the proteasome. It also inhibits tumor cell growth, induces cell cycle arrest and apoptosis, and suppresses tumor immune evasion.
  • Degrades STAT3 transcription factor.
  • Exhibits anti-tumor activity in HeLa and MCF-7 cells.
  • Recruits E3 ligase VHL for STAT3 degradation.
  • Inhibits tumor cell growth.
  • Induces cell cycle arrest and apoptosis.
  • Suppresses tumor immune evasion.
Catalog No. 50-003-85373