Learn More
Medchemexpress LLC STAT3-IN-3 | 2361304-26-7 | 98.4% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128588100MG
Encompass_Preferred
STAT3-IN-3 is a potent and selective inhibitor of signal transducer and activator of transcription 3 (STAT3). It exhibits anti-proliferative activity by inducing apoptosis in breast cancer cells and is considered a promising mitochondria-targeting STAT3 inhibitor for cancer research. This compound inhibits STAT3 tyrosine and serine phosphorylation and its DNA-binding activity. In vivo studies have shown potent antitumor activity against implanted 4T1 breast tumors.
  • Potent and selective STAT3 inhibitor
  • Induces apoptosis in breast cancer cells
  • Mitochondria-targeting for cancer research
  • Inhibits STAT3 phosphorylation and DNA-binding
  • Down-regulates STAT3 target genes
  • Increases ROS production
  • Reduces mitochondrial membrane potential
  • Induces caspase cleavage
  • Exhibits anti-proliferative activity
  • Potent antitumor activity in vivo
Catalog No. 50-003-41884