Learn More
Medchemexpress LLC HY-128588 5mg Medchemexpress, STAT3-IN-3 CAS:2361304-26-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285885MG
Encompass_Preferred
Medchemexpress, HY-128588 5mg STAT3-IN-3 CAS:2361304-26-7 STAT3-IN-3 is a potent and selective inhibitor of signal transducer and activator of transcription 3 (STAT3). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9754